Learn More
HELB Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24700-1-AP-20UL
This item is not returnable.
View return policy
Description
HELB gene ontology annotations related to this gene include cellular response to DNA damage stimulus; DNA replication, synthesis of RNA primer; negative regulation of double-strand break repair via homologous recombination; regulation of DNA double-strand break processing.Specifications
| HELB | |
| Polyclonal | |
| Unconjugated | |
| HELB | |
| DNA helicase B, hDHB, HELB, helicase (DNA) B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 92797 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8NG08 | |
| HELB | |
| HELB Fusion Protein Ag20335 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |