Learn More
Heidolph™ Hei-VAP Low Boiling Dry Ice Package, EU-plug
Shop All Heidolph Instruments Products
Click to view available options
Plug Type:
EU-plug
Unit Size:
Each
Description
- Hei-VAP Core with motor lift and coated G5 dry ice condenser
- Vacuum pump Rotavac Valve Tec
- Manual vacuum controller
- Tubing set
Specifications
Specifications
| Type | Motor Lift Rotary Evaporator |
| Plug Type | EU-plug |