Learn More
Heidolph™ Hei-VAP Industrial B Rotary Evaporator with Glassware Set
Shop All Heidolph Instruments Products
Click to view available options
For Use With (Application):
All Standard Distillations
Hood applications or where height is restricted
Low-boiling solvents - cooling by dry-ice
Low-boiling solvents - highest recovery rates
Solutions that tend to splash and foam
Glassware Assembly Type:
Set A
Set A2
Set A2C
Set AC
Set K
Set KC
Set R
Set RC
Set Shorty R
Set Shorty RC
Unit Size:
Each
Description
- Universal heating bath accommodates water or other bath fluids allowing for temperature settings up to 180°C
- Receiver cassettes protect against threat of glassware breakage
- Comes as standard with integrated refill water system, spillover prevention and a release valve at the bottom
- Optional Base Cart is fully-maneuverable with removable panels for storage of vacuum pump, glassware, solvents or accessories
- Certification according to GMP available for this model: validation for installation (IQ) and operating qualification (OQ)
- Large touch screen control panel with illuminated displays for all process parameters, programmable ramps
Features identical specifications as the Hei-VAP Industrial, but for limited budgets and comes without these safety aspects:
- Metal frame and safety glass guard hood covering the bath
- High-impact PMMA door covering the glassware assembly
- High-impact PMMA latching doors which cover the receiving flask cassettes
Specifications
Specifications
| Speed | 6 to 160 rpm |
| Temperature Range (Metric) | 20°C to 180°C |
| Display | 7 in. touchpanel |
| For Use With (Application) | Hood applications or where height is restricted |
| Includes | 1 ascending condenser (320 mm shorter than traditional one), 1 20 L evaporating flask, (2) 10 L receiving flasks |
| Glassware Assembly Type | Set Shorty RC |
| IP Rating | IP20 |
| Temperature Accuracy | ±1°C |
| Heating Capacity (Metric) | 4000 W |
| Type | Glassware Assembly Rotary Evaporator |
| Show More |