Learn More
Heidolph™ Hei-PLATE Sensor Basic Package Ultimate
Product Code. 17981971 Shop All Heidolph Instruments Products
Change view
Click to view available options
Plug Type:
EU
Size:
135 mm dia.
145 mm dia.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type Size Unit Size
17981971 EU 135 mm dia. Each
17941971 EU 145 mm dia. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17981971 Supplier Heidolph™ Supplier No. 5064215000
  • ONLINE EXCLUSIVE PRICE
    21220.00 NOK
    18390.40 NOK / Pack of 1
    Save 2829.60 NOK
    13% Off
View all promotions
Estimated Shipment: 04-12-2026
Hei-PLATE Sensor Basic Package Ultimate includes everything needed for fast and precise heating.
  • Magnetic stirrer Hei-PLATE Mix'n'Heat Ultimate provides fast and precise heating without overshoot
  • For sample-friendly work
  • Pt1000 temperature sensor for precise temperature regulation
TRUSTED_SUSTAINABILITY
Specifications
Top Plate Material Ceramic-coated Aluminum
Voltage 230 V
Size 135 mm dia.
Temperature (Metric) 20°C to 300°C
Type Magnetic Stirring Hotplate
Shape Round
Max. Sample Load 25 kg
Plug Type EU
No. of Positions 1
Max. Container Size 20 L
Stirring Range 100 to 1400 rpm
Warranty 3 Years
Certifications/Compliance CE, NRTL
Display TFT