Description
- Magnetic stirrer Hei-PLATE Mix'n'Heat Expert provides fast and precise heating without overshoot
- Gentle to the sample
- Pt1000 - enables precise temperature regulation
- 1 L Heat-On Blocks ensure optimal heat transfer without the use of water or oil
- Application tip: Fill the flask (not in the scope of delivery) only halfway to avoid loss of solvent
- Findenser™ completes the system as a high-performance condenser that is used without the use of water (Performance is optimal for solvent volumes up to 1 L)
Specifications
Specifications
| Top Plate Material | Ceramic-coated Aluminum |
| Voltage | 230 V |
| Size | 145 mm dia. |
| Temperature (Metric) | 20°C to 300°C |
| Type | Magnetic Stirring Hotplate |
| Shape | Round |
| Max. Sample Load | 25 kg |
| Plug Type | EU |
| No. of Positions | 1 |
| Max. Container Size | 20 L |
| Show More |