Learn More
Heidolph™ Hei-FLOW Ultimate 120
Product Code. 10384721 Shop All Heidolph Instruments Products
Change view
Click to view available options
Motor Speed:
5 to 120 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Motor Speed Unit Size
10384721 5 to 120 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10384721 Supplier Heidolph™ Supplier No. 5235201000
  • ONLINE EXCLUSIVE PRICE
    45685.00 NOK
    39598.31 NOK / Pack of 1
    Save 6086.69 NOK
    13% Off
View all promotions
Estimated Shipment: 04-12-2026
With the Hei-FLOW Ultimate 120, you can choose between a single-channel or multichannel pump head.
  • Accurate dosing is particularly important in the pharmaceutical, cosmetic or food sector. Comprehensible process conditions are a prerequisite for the safe conveyance of sensitive media. This is ensured by a control accuracy of ±1% and precise speed adjustment (5 to 120 rpm).
  • In addition, an optional foot switch allows the pump to be operated in closed exhaust hoods. The parameters - speed, direction of rotation and on/off function - can be controlled externally either through the analog or digital interface.
  • In this model, you can choose between a single-channel or multichannel pump head. Flow rates vary depending on the pump head: the single-channel head has a flow rate of 0.38 to 813 mL/min.; for the multi-channel head 0.005 to 364 mL/min. Depending on the focus of the process, the following parameters can be set on the device: Speed, tubing diameter, flow rate display, metering volume, interval dosing and break times. The pump can be calibrated for optimum dosing results.
TRUSTED_SUSTAINABILITY
Specifications
Accuracy ±1%
Control Type Digital Variable Speed
Display Type LED
Enclosure Rating IP33
Flow Rate 0.38 to 813 mL/min. (Single-channel), 0.005 to 364 mL/min. (Multi-channel)
Motor Speed 5 to 120 rpm
Voltage 230 V
Plug Type EU Plug