Learn More
Heidolph™ Hei-Flow Precision 01 Multi-Channel Peristaltic Pump
Product Code. 10534552 Shop All Heidolph Instruments Products
Change view
Click to view available options
Flow Rate:
0.005 to 364 mL/min.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Unit Size
10534552 0.005 to 364 mL/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10534552 Supplier Heidolph™ Supplier No. 5235201300
  • ONLINE EXCLUSIVE PRICE
    47255.00 NOK
    40960.96 NOK / Pack of 1
    Save 6294.04 NOK
    13% Off
View all promotions
Estimated Shipment: 12-01-2027
Includes leading safety standards and features for superior ease of use and reduced cost of ownership. Heidolph™ Hei-Flow Precision 01 Multi-Channel Peristaltic Pump includes a variety of features to ensure easy control and accurate flow rate.
  • Control speed (rpm), direction and on/off function via analog or digital interface.
  • Flow characteristic of pump heads in combination with various tubing diameters is pre-programmed for accurate flow rate numbers.
  • Digital read-out of:
    • Speed (rpm).
    • Tubing diameter.
    • Flow rate in mL/min.
    • Dosing volume.
    • Interval dosing.
    • Pause function.
  • Easily change all process parameters via control buttons.
  • Connect to an optional foot-pedal remote control via cable to start and stop your pump drive outside a closed fume hood to increase your personal safety and provide ease of use for multiple sample fills.
  • Electronic speed control with ± 0.5 % accuracy.
  • Max button to accelerate tube filling and draining.
  • Clockwise or counter-clockwise operation at the press of a button.
  • Individual calibration of flow volume and flow rate.
  • Suitable for multi-channel operation.
  • Upgradable in minutes with a pump head adaptor.
TRUSTED_SUSTAINABILITY
Specifications
Flow Rate 0.005 to 364 mL/min.
Motor Speed 5 to 120 rpm