Learn More
Heidolph™ Hei-Flow Peristaltic Pump Silver 1 Package.
Product Code. 10192562 Shop All Heidolph Instruments Products
Change view
Click to view available options
Flow Rate:
0.83 to 861 mL/min.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Unit Size
10192562 0.83 to 861 mL/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10192562 Supplier Heidolph™ Supplier No. 5235001900
  • ONLINE EXCLUSIVE PRICE
    42035.00 NOK
    40922.33 NOK / Pack of 1
    Save 1112.67 NOK
    2% Off
Estimated Shipment: 07-12-2026
Satisfy a variety of peristaltic pump needs with this package kit. Heidolph™ Hei-Flow Peristaltic Pump Silver 1 Package includes quality accessory components.
  • Includes pump, pump head, and tubing.
TRUSTED_SUSTAINABILITY
Specifications
Description Single channel, fits 1.6 mm tubing
Flow Rate 0.83 to 861 mL/min.
Motor Speed 10 to 120 rpm
Includes SP quick pump head, Hei-FLOW Value 01 pump, 1 m each Tygon (standard) and Silicone tubing (inner diameter:3.1 mm).