Learn More
Heidolph™ Hei-FLOW Expert 120
Product Code. 10045731 Shop All Heidolph Instruments Products
Change view
Click to view available options
Motor Speed:
5 to 120 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Motor Speed Unit Size
10045731 5 to 120 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10045731 Supplier Heidolph™ Supplier No. 5235101000
  • ONLINE EXCLUSIVE PRICE
    37210.00 NOK
    32252.41 NOK / Pack of 1
    Save 4957.59 NOK
    13% Off
Estimated Shipment: 24-11-2026
The pump drive of the model 01 of the Hei-FLOW Advantage series can be operated with either single-channel or multi-channel pump heads.
  • The pump drive of the model 01 of the Hei-FLOW Advantage series can be operated with either single-channel or multi-channel pump heads. The flow rates with single-channel pump heads are in the range of 0.38 to 813 mL/min; with multi-channel pump heads from 0.005 to 329 mL/min.
  • By integrating an analog interface, it is possible to control all functions externally. In addition, an optional foot switch can be adapted to enable dosing by foot pressure. Hands thus remain free to perform other work, for example, to hold the tubing and the vessel.
  • A speed of 5 to 120 rpm with a control accuracy of ±3.5% guarantees optimum process results.
TRUSTED_SUSTAINABILITY
Specifications
Accuracy ±3.5%
Control Type Digital Variable Speed
Display Type LED
Enclosure Rating IP33
Flow Rate 0.38 to 813 mL/min. (Single-channel), 0.005 to 329 mL/min. (Multi-channel)
Motor Speed 5 to 120 rpm
Voltage 230 V
Plug Type EU Plug