Learn More
Heidolph™ Hei-FLOW Core 120 Multi
Product Code. 10321302 Shop All Heidolph Instruments Products
Change view
Click to view available options
Motor Speed:
10 to 120 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Motor Speed Unit Size
10321302 10 to 120 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10321302 Supplier Heidolph™ Supplier No. 5235001300
  • ONLINE EXCLUSIVE PRICE
    35960.00 NOK
    31166.33 NOK / Pack of 1
    Save 4793.67 NOK
    13% Off
View all promotions
Estimated Shipment: 24-11-2026
This peristaltic pump is ideal for the simple conveying of all media.
  • The pump drive of this model makes it possible to switch between a single channel and multi-channel pump head. By simply flipping the switch, it can also be conveyed in an anti-clockwise direction.
  • Depending on the adapted pump head, a flow rate of 0.83 to 861 mL/min for the single-channel and 0.005 to 364 mL/min for the multi-channel pump head is guaranteed. The Value 01 conveys with a control accuracy of ± 5% at a speed range of 10 to 120 rpm.
TRUSTED_SUSTAINABILITY
Specifications
Accuracy ± 5%
Control Type Digital Variable Speed
Display Type LED
Enclosure Rating IP33
Flow Rate 0.83 to 861 mL/min (Single-channel), 0.005 to 364 mL/min. (Multi-channel)
Motor Speed 10 to 120 rpm
Voltage 230 V
Plug Type EU Plug