Learn More
Heidolph™ Hei-Connect
Shop All Heidolph Instruments Products
Click to view available options
Plug Type:
EU
Unit Size:
Each
Description
- The Hei-Connect combines gentle stirring (up to 1400 rpm) with a precise heating function (up to 300°C).
- Unique in its robust design and secure functionality, this model stands out with additional features giving you complete control and an easy, accurate documentation of your process parameters.
- The unique Kera-Disk™ footprint chemically resistant, scratch-proof, and has a diameter of 145 mm.
- This unique size makes it suitable for use with many accessories and samples on a small footprint.
- The anodized aluminum from which it is made enables the fastest possible heating times.
- The Hei-Connect magnetic stirrer provides a digital display for setting your desired temperature exactly to the degree.
- This precise temperature without overshooting is controlled by two selectable heating modes (FAST/PRECISE) and the optional external temperature sensor Pt1000.
- To facilitate process control and documentation, the Hei-Connect is equipped with a timer and an RS 232 interface.
- The free Hei-Control Software is included in the scope of delivery.
Specifications
Specifications
| Top Plate Material | Aluminum Alloy |
| Voltage | 230 V |
| Temperature (Metric) | 300°C |
| Temperature (English) | 572°F |
| Frequency | 50/60 Hz |
| Type | Magnetic Stirring Hotplate |
| Shape | Round |
| Plug Type | EU |
| No. of Positions | Multiple |
| Display | Digital |
| Show More |