missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HECTD4 Monoclonal Antibody (A2-F5-F10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA532878
This item is not returnable.
View return policy
Description
HECTD4 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
| HECTD4 | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.2% BSA, 50% Glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9Y4D8 | |
| HECTD4 | |
| Recombinant protein within Human Protein C12orf51 aa 232-516. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| A2-F5-F10 | |
| Unconjugated | |
| HECTD4 | |
| A130010L24; AF-1 specific protein phosphatase; AU042671; BC019639; C12orf51; EG545802; Gm15800; HECT domain containing E3 ubiquitin protein ligase 4; HECT domain E3 ubiquitin protein ligase 4; HECT domain-containing protein 4; HECTD4; mKIAA0614; OTTMUSG00000026283; POTAGE; predicted gene 15800; probable E3 ubiquitin-protein ligase C12orf51; probable E3 ubiquitin-protein ligase HECTD4; transmembrane protein C12orf51 | |
| Mouse | |
| Protein A | |
| RUO | |
| 269700, 283450 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction