Learn More
Cole-Parmer™ Heavy Duty PS Pinch Clamp Closure for Tubing and Hose
Handle higher working pressures with these heavy-duty tubing closures. Cole-Parmer™ Heavy Duty Closures provide excellent durability, can be continually reused and resist most chemicals.
Brand:  Cole-Parmer™ 20001-00
Product Code. 15394677
  • 742.00 NOK / Each
Only null left
Estimated Shipment: 20-02-2026
Description
  • High-strength polysulfone clamp
  • Quick installation with positive  swing-away locking screw
  • Smooth contours allow for easy cleaning
  • Simply align the hose within the clamp; mating surfaces on the clamp indicate complete closure 
  • Typical applications include sterile filling, media bag connections, pumping, liquid control and high-purity fluid transfers
  • FDA-compliant polysulfone material can be sterilized by autoclave or gamma radiation