Learn More
Heidolph™ Heating Module
Description
- Even and fast heat distribution is essential for outstanding results
- With an output of 300 watts, the heating module ensures fast heating times within the incubation system
- Thermophilic organisms can also be incubated at a temperature of 65°C
Specifications
Specifications
| Voltage | 115/230 V |
| Temperature Range (Metric) | 5°C to 65°C |
| For Use With (Equipment) | Incubator 1000 |