Learn More
Bibby Scientific™ Heating Electromantle
Product Code. 11708422
Change view
Click to view available options
Capacity (Metric):
250 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11708422 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11708422 Supplier Bibby Scientific™ Supplier No. 3610465
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Capacity (Metric) 250 mL
For Use With EMV0250/CEX1
Shape V-shaped
Product Type Heating Electromantle
Temperature (Metric) 450°C