Learn More
Grant Instruments™ Heating Block for TS-DW Deep-well Microplate Thermoshaker
Product Code. 15440474
Click to view available options
Holds:
Axygen 96 Deep-well Plate
Eppendorf 96 Deep-well Plate
Sarstedt 96 Deep-well Plate
Starlab 96 Deep-well Plate
Size:
1000 μL
1200 μL
2200 μL
Unit Size:
Each
Product Code. 15440474
Brand: Grant Instruments™ B2S
Only null left
Add to basket
Add to basket
Accommodates one deep-well plate for TS-DW Deep-well Microplate Thermoshaker
Block for one deep-well plate
  • Shaped to fit the microplates for better heat transfer
  • Three year warranty
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) TS-DW Microplate Thermoshaker
Holds Sarstedt 96 Deep-well Plate
Type Block
Size 2200 μL