Learn More
IKA™ Heating Block
Product Code. 17593636 Shop All IKA Products
Change view
Click to view available options
For Use With (Equipment):
Magnetic Stirrer RCT Digital
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
17593636 Magnetic Stirrer RCT Digital Each
1 options
Product Code. 17593636
Brand: IKA™ 0020103369
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
IKA Flow Heating Block holds four disc reactors which are fed by the IKA FLOW during reaction.
  • IKA Flow Heating Block is installed to the magnetic stirrer RCT digital and has the maximum capacity of holding four disc reactors which are fed by the IKA FLOW during reaction.
  • Disc reactos can be purchased separately.
  • Scope of delivery
    • Heating Block
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Magnetic Stirrer RCT Digital