Learn More
Labconco™ Heated Product Shelves, Small
Description
Heated Product Shelves consist of three aluminum shelves with 1/4 in. rims, three stainless steel support rods, nine clips for shelf height adjustment, low voltage transformer with variable heat control to +40° C (+104° F), and three rubber feet. Product Shelves accommodate bulk samples, microtiter plates, vials or serum bottles. They are designed to fit inside Drying Chambers (sold separately).
Specifications
Specifications
| Type | Heated Shelf |
| Material | Aluminum |
| Certifications/Compliance | CE |
| For Use With (Equipment) | Drying Chambers |
| Voltage | 0 to 300 V |