Learn More
Brady™ PermaSleeve™ Heat-shrink Polyolefin Wiremarker Sleeves
Product Code. 11791765
Click to view available options
Color:
Black on Yellow
Yellow
Wire Gauge:
16 AWG - 8 AWG
20 AWG - 10 AWG
Dimensions (LxW):
2.13 m x 11.15 mm
42.16 x 25.78 mm
Quantity:
1 Ea.
1000 Pk.
Unit Size:
Each
Product Code. 11791765
Brand: Brady™ 115013
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Bulk roll of PermaSleeve markers. PermaSleeve™ Heat-shrink Polyolefin Wiremarker Sleeves provide a wire marking solution that combines speed, ease of use, and permanence to create long-lasting, high quality identification.
  • These Sleeves are self-extinguishing.
  • Meet the requirements of SAE AMS-DTL-23053/5 Class 1, SAE-AS-8153, MIL-STD-202 method 215K and is UL224 recognised.
  • CSA acceptance pending.
Specifications
Certifications/Compliance ASTM D 2671, ASTM E 162, ASTM E 662, ASTM G 155, MIL-STD-202, SAE-AMS-DTL-23053, SAE-AS-81531, UL Recognised
Description Bulk PermaSleeve Wire Marker Sleeves for BMP61 and BMP71
Color Yellow
Finish Matte
Format Roll
Material Polyolefin
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Wire and Cable Markers
For Use With (Application) Electrical labelling, Wire and cable labelling
For Use With (Equipment) BMP61, BMP71
Labels per Row 1
Min. Operating Temperature (Metric) -55°C
No. per Pack 1000 Sleeves/Box
Quantity 1000 Pk.
Shape Rectangle
Shrink Ratio 3:1
Temperature Range (English) -67°F to +275°F
Temperature Range (Metric) -55°C to +135°C
Vertical Repeat (Metric) 19.05 mm
Wire Gauge 20 AWG - 10 AWG
Dimensions (LxW) 42.16 x 25.78 mm
Width (English) 1 in.
Width (Metric) 25.4 mm
Width (English) Printable Area 1 in.
Height (English) 0.34 in.
Height (Metric) 8.51 mm
Diameter (English) 0.06 to 0.15 in.
Diameter (Metric) 1.57 to 3.81 mm