Learn More
Epredia™ Heat-Sealable Polyethylene Bags, 3 x 5 in. (7.6 x 12.7cm)
Product Code. p-4525099
Click to view available options
Length (English):
9 in.
Unit Size:
Pack of 100
Product Code. 12734878
Brand: Epredia™ 844
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Protect specimens for mailing with Epredia™ Heat-Sealable Polyethylene Bags.
  • Heavy-duty polyethylene construction
  • Heat-sealable to ensure leak-resistant storage
Ordering Information:
  • Sold in packs of 100
TRUSTED_SUSTAINABILITY
Specifications
Product Type Heat-Sealable Bag
Material Polyethylene
Diameter (Metric) 240 mm
Length (English) 9 in.
Length (Metric) 22.9 cm
Width (English) 16 in.
Width (Metric) 40.6 cm