Learn More
Keystone Cleanroom Products Heat Seal Tyvek™ Steam Sterilization Pouches
BIOPHARMAGREENER_CHOICE
Product Code. 17757817 Shop All Keystone Cleanroom Products Products
Click to view available options
Dimensions (L x W):
3 x 8 in.
6 x 3 in.
8 x 6 in.
10 x 8 in.
12 x 6 in.
15 x 8 in.
4 x 15 in.
6 x 25 in.
12 x 12 in.
17 x 12 in.
17 x 16 in.
18 x 12 in.
18 x 24 in.
20 x 10 in.
22 x 18 in.
24 x 12 in.
24 x 24 in.
30 x 10 in.
30 x 24 in.
Unit Size:
Pack of 1000
Pack of 250
Pack of 500
Product Code. 17757817
Brand: Keystone Cleanroom Products BBMGTVKHS1617
Heat Seal Tyvek™ Steam Sterilization Pouches provide a strong barrier from microbial contamination.
  • Great choice for equipment parts that are heavy, have sharp edges, or packaging that requires surface sanitization post-sterilization
  • DuPont Tyvek™/Polyester-based flexible film is lint-free, moisture and puncture resistant, extremely tough, and designed to be autoclaved
  • The Tyvek™ side offers high breathability while providing a strong barrier from microbial contamination
  • The clear film side offers high clarity along with strength and durability
  • Should be heat sealed closed using a validate-able impulse, or continuous band heat sealer
  • Each pouch has a CLASS A process indicator, which changes from pink to dark brown when exposed to steam
  • When the peel-able seals are opened, virtually no airborne particulates are generated
  • Recommended for sterilization of pharmaceutical equipment
  • Manufactured and packaged in controlled cleanroom environment
  • Works with both Impulse and Continuous Band Sealers
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance (COC)
Closure Type Heat Seal
Color White
Disposable Single-use
For Use With (Application) Steam Sterilization
Material Tyvek
Sterility Non-sterile
Latex Free Yes
Quantity 500/Pk.
Dimensions (L x W) 17 x 16 in.
Length (English) 17 in.
Length (Metric) 43.2 cm
Width (English) 16 in.
Width (Metric) 40.6 cm
Product Type Autoclave Bag