Learn More
Heidolph™ Insert for Multi-Well Holders
Product Code. 10237733 Shop All Heidolph Instruments Products
Change view
Click to view available options
Type:
100mL Insert
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10237733 100mL Insert Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10237733 Supplier Heidolph™ Supplier No. 5058006400
  • ONLINE EXCLUSIVE PRICE
    1715.00 NOK
    1674.94 NOK / Pack of 1
    Save 40.06 NOK
    2% Off
Estimated Shipment: 23-11-2026
Insert for the unique Multi-Well Holder that is designed to hold either one or two inserts for flasks that also features cut-away sides for use with two or three neck flasks and accept the optional safety lifting handles.
Insert for the Heat-On 100-ml Multi-Well Holder.
TRUSTED_SUSTAINABILITY
Specifications
Type 100mL Insert