Learn More
Grant Instruments™ Heat Exchange Cooling Coil
Product Code. p-8328236
Change view
Click to view available options
For Use With (Equipment):
OLS26
T100, TC120, TX150, TXF200
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
15345872 OLS26 Each
11486487 T100, TC120, TX150, TXF200 Each
2 options
Product Code. 15345872
Brand: Grant Instruments™ CW26
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Can be attached to a cold water supply or refrigerated circulator. Grant Instruments™ Heat Exchange Coil maintains bath down to 2°C above the temperature of the coolant.
  • Stainless steel heat exchange coil that can be attached to cold water supply or a refrigerated circulator
  • Can be used to maintain a water bath temperature that is 2°C above the temperature of the coolant used
  • Compatible with OLS 26 Aqua Pro
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) OLS26
Type Heat Exchange Coil