Learn More
R&D Systems™ Heart Tissue Slides (Adult Normal)- Paraffin
Product Code. 18113079 Shop All Bio Techne Products
Change view
Click to view available options
5 Slides
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18113079 5 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18113079 Supplier R&D Systems™ Supplier No. NBP246893
  • ONLINE EXCLUSIVE PRICE
    3875.00 NOK
    3662.40 NOK / Pack of 5
    Save 212.60 NOK
    5% Off
 Restricted Item
Estimated Shipment: 16-11-2026
A Novus product, part of R&D Systems. Useful in immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Description 50 Cores: 59 carcinoma, adenocarcinoma, carcinosarcoma
Quantity 5 Slides
Tissue Condition Normal
Type Heart
Dimensions (LxW) 2.0 mm
Product Line Novus