Learn More
Masterflex™ HDPE Y Connector
Product Code. p-8012995
Click to view available options
For Use With:
4.76mm
9.52mm
12.7mm
Unit Size:
Pack of 10
Product Code. 12324598
Brand: Masterflex™ 3061421
Only null left
Add to basket
Add to basket
Designed to tight tolerances, these fittings match perfectly to your tubing to ensure smooth flow and a proper fit.
  • Designed to tight tolerances
  • Made from virgin materials
  • Very good chemical resistance
  • Temp range: -53 to 65°C
  • Sterilize by ethylene oxide only
Specifications
Diameter (Metric) Inner 12.7 mm
Material HDPE
For Use With 12.7mm
Min. Operating Temperature (Metric) -53°C
Packaging Quantity 10 Pack
Max. Operating Temperature (Metric) 65°C
Product Type Y Connector