Learn More
Masterflex™ HDPE Y Connector
Description
- Designed to tight tolerances
- Made from virgin materials
- Very good chemical resistance
- Temp range: -53 to 65°C
- Sterilize by ethylene oxide only
Specifications
Specifications
| Diameter (Metric) Inner | 9.52 mm |
| Material | HDPE |
| For Use With | 9.52mm |
| Min. Operating Temperature (Metric) | -53°C |
| Packaging Quantity | 10 Pack |
| Max. Operating Temperature (Metric) | 65°C |
| Product Type | Y Connector |