Learn More
BRAND™ HDPE Wide Neck Storage Bottles
Product Code. p-8546001 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
5 L
10 L
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10319811 5 L Each
10004321 10 L Each
2 options
Product Code. 10319811
Brand: BRAND™ 130970
Only null left
Add to basket
Add to basket
HDPE construction. Wide neck bottle includes screw cap and carrying handle.
  • HDPE construction
  • Wide neck
  • Includes screw cap and carrying handle
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 5 L
Includes Carrying handle
Material HDPE
Closure Included Yes
Product Type Storage Bottle
Closure Type Screw Cap