Learn More
Buerkle™ HDPE Wide Mouth Disposal Keg Container
Product Code. p-8002253 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
3.6 L
6 L
10 L
26 L
34 L
Unit Size:
Each
Product Code. 10427981
Brand: Buerkle™ 11050026
Only null left
Add to basket
Add to basket
HDPE Wide Mouth Disposal Keg Container for disposal or storage of powders, granulates and liquids. Can be closed tightly and sealed. The containers have an UN transport approval for solids.
  • Made of HDPE
  • Temperature resistant from -20 °C to +80 °C
  • UN transport approval for solids 1H2/X
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 26 L
Description Wide mouth disposal keg
For Use With (Application) Used for Waste Disposal or Storage of Liquids, Powders or Granules
Includes Cap
Material HDPE
Certifications/Compliance UN transport approval 1H2/X
Shape Round
Height (Metric) 425 mm
Diameter (Metric) 316 mm