Learn More
Buerkle™ HDPE Wide Mouth Disposal Keg Container
Product Code. 10488331 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
3.6 L
6 L
10 L
26 L
34 L
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10488331 10 L Each
10427981 26 L Each
15445554 3.6 L Each
10593242 6 L Each
10468222 34 L Each
5 options
Product Code. 10488331
Brand: Buerkle™ 11050010
Only null left
Add to basket
Add to basket
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
HDPE Wide Mouth Disposal Keg Container for disposal or storage of powders, granulates and liquids. Can be closed tightly and sealed. The containers have an UN transport approval for solids.
  • Made of HDPE
  • Temperature resistant from -20 °C to +80 °C
  • UN transport approval for solids 1H2/X
TRUSTED_SUSTAINABILITY
Spezifikation
Capacity (Metric) 10 L
Description Wide mouth container
Material HDPE
Height (Metric) 238 mm
Diameter (Metric) 274 mm