Learn More
Masterflex™ HDPE T-Connector Barbed Fittings
Click to view available options
For Use With:
1/4 in. tubing I.D.
3/16 in. tubing I.D.
3/4 in. tubing I.D.
3/8 in. tubing I.D.
5/16 in. tubing I.D.
Unit Size:
Pack of 10
Description
- Use with low-pressure applications, generally less than 30 psi (2 bar)
- Manufactured from quality HDPE
- Sterilization by ethylene oxide (ETO) only
- Complete range of standard sizes available
- Provided in packs of 10
Specifications
Specifications
| Material | HDPE |
| For Use With | 3/16 in. tubing I.D. |
| Packaging Quantity | 10 Pack |
| Sterilization Method | Ethylene Oxide |
| Shape | T-shape |
| Product Type | Barbed Fitting |