Learn More
Masterflex™ HDPE T Connector
Click to view available options
For Use With:
4.76mm
9.52mm
15.8mm
19.05mm
25.4mm
Unit Size:
Pack of 10
Specifications
Specifications
| Diameter (Metric) Inner | 25.4 mm |
| Material | HDPE |
| For Use With | 25.4mm |
| Min. Operating Temperature (Metric) | -53°C |
| Packaging Quantity | 10 Pack |
| Shape | T |
| Max. Operating Temperature (Metric) | 65°C |
| Product Type | T-Connector |