Learn More
Masterflex™ HDPE T Connector
Product Code. 12344588
Click to view available options
For Use With:
4.76mm
9.52mm
15.8mm
19.05mm
25.4mm
Unit Size:
Pack of 10
Product Code. 12344588
Brand: Masterflex™ 3061325
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Diameter (Metric) Inner 15.8 mm
Material HDPE
For Use With 15.8mm
Min. Operating Temperature (Metric) -53°C
Packaging Quantity 10 Pack
Shape T
Max. Operating Temperature (Metric) 65°C
Product Type T-Connector