Learn More
Cole-Parmer™ HDPE Stirring Paddle
Product Code. 15346181 Shop All Cole-Parmer Products
Click to view available options
Length (English):
72 in.
Unit Size:
Each
Product Code. 15346181
Brand: Cole-Parmer™ 0626674
Only null left
Add to basket
Add to basket
Ideal for mixing or taking samples from large tanks or vats
  • 72-inch Long Stirring Paddle
  • Larger size paddles and scrapers for mixing or taking samples from large tanks or vats
  • Stirring paddles are made of high-density polyethylene (HDPE)
  • Nylon scraping paddles will not scratch glass
Specifications
Product Type Stirring Paddle
Length (English) Blade 6 in.
Length (English) 72 in.
Width (English) Blade 3 in.
Material HDPE