Learn More
Ratiolab™ HDPE Pasteur Pipet
Description
- No risk of cross-contamination or carry-over
- No danger of glass breakage with the resulting danger of infectious material escaping
- Transparent polyethylene construction
- Inert to biological fluids and most acids
- Soft, elastic bulb
- Burr-free orifice for neat, clean and precise dispensing
- Clearly visible embossed graduations
- BSE-free
- Can be used for food
Specifications
Specifications
| Material | HDPE |
| Sterility | Non-sterile |
| For Use With (Application) | Food analysis |
| Quantity | 400 Pack |
| Volume (Metric) | 6 mL |
| Length (Metric) | 148 mm |
| Type | Pasteur Pipette |