Learn More
Buerkle™ HDPE Narrow Necked Square Bottle with Screw Cap
Product Code. p-8658016 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
100 mL
1000 mL
Unit Size:
Each
Product Code. 15495574
Brand: Buerkle™ 03320100
Only null left
Add to basket
Add to basket
Buerkle™ HDPE Narrow Necked Square Bottle with Screw Cap is a cost-effective bottle for packing and storing of liquids.
  • HDPE, transparent, shatterproof
  • Special labelling area for labels or manual lettering
  • Suitable as a disposable bottle
  • Leakproof
  • With screw cap
  • Dropper cap 0300-3018 is available but sold separately
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 100 mL
Color Transparent
Description Excludes dropper cap 0300-3018
Dimensions (L x W x H) 41 x 33 mm
Diameter (Metric) Inner Neck 13 mm
For Use With (Application) Foodstuffs
Includes Screw cap
Material HDPE
Length (Metric) 41 mm
Quantity 1
Width (Metric) 33 mm
Thread Size (Metric) 18 mm
Closure Color Transparent
Closure Included Yes
Mouth Narrow
Shape Square
Height (Metric) 113 mm
Product Type Square Narrow Mouth Bottle
Closure Material HDPE
Closure Type Screw Cap