Learn More
Masterflex™ HDPE Male Pipe Adapter Barbed Fitting
Product Code. 11725489
Change view
Click to view available options
For Use With:
1/2 NPT(M) x 1/2 in. tubing I.D.
1/2 NPT(M) x 3/8 in. tubing I.D.
1/4 NPT(M) x 1/4 in. tubing I.D.
1/8 NPT(M) x 3/8 in. tubing I.D.
3/4 NPT(M) x 1/4 in. tubing I.D.
3/4 NPT(M) x 3/8 in. tubing I.D.
3/4 NPT(M) x 5/8 in. tubing I.D.
3/8 NPT(M) x 1/2 in. tubing I.D.
3/8 NPT(M) x 1/4 in. tubing I.D.
3/8 NPT(M) x 3/8 in. ID Tubing
Thread Size:
1/2 NPT(M)
1/4 NPT(M)
1/8 NPT(M)
3/4 NPT(M)
3/8 NPT(M)
Unit Size:
Pack of 10
Product Code. For Use With Thread Size unitSize
11725489 3/8 NPT(M) x 1/2 in. tubing I.D. 3/8 NPT(M) Pack of 10
11705549 3/4 NPT(M) x 1/4 in. tubing I.D. 3/4 NPT(M) Pack of 10
11715489 3/8 NPT(M) x 1/4 in. tubing I.D. 3/8 NPT(M) Pack of 10
11755259 1/2 NPT(M) x 1/2 in. tubing I.D. 1/2 NPT(M) Pack of 10
11745489 1/2 NPT(M) x 3/8 in. tubing I.D. 1/2 NPT(M) Pack of 10
11765708 1/8 NPT(M) x 3/8 in. tubing I.D. 1/8 NPT(M) Pack of 10
11785479 1/4 NPT(M) x 1/4 in. tubing I.D. 1/4 NPT(M) Pack of 10
11795718 3/4 NPT(M) x 3/8 in. tubing I.D. 3/4 NPT(M) Pack of 10
11755708 3/8 NPT(M) x 3/8 in. ID Tubing 3/8 NPT(M) Pack of 10
11799998 3/4 NPT(M) x 5/8 in. tubing I.D. 3/4 NPT(M) Pack of 10
10 options
Product Code. 11725489
Brand: Masterflex™ 3062164
Only null left
Add to basket
Add to basket
For quick and easy connection of fluid paths in the laboratory without the use of tools.
  • Slip flexible tubing or hose onto the barb to create a seal that typically holds pressures up to 30 psi (2 bar)
  • Available in a variety of configurations to meet your needs
  • Adapts from a NPT male thread to a hose barb
  • Sterilization through ethylyene oxide only
  • Provided in packs of 10
Specifications
Material HDPE
For Use With 3/8 NPT(M) x 1/2 in. tubing I.D.
Packaging Quantity 10 Pack
Sterilization Method Ethylene Oxide
Thread Size 3/8 NPT(M)
Shape Straight
Product Type Barbed Fitting