Learn More
Gosselin™ HDPE, Jerry Can (Sterile by Irradiation)
Product Code. 11733114
Change view
Click to view available options
:
Capacity: 2.5L; 220L x 160W x 105mmH
Product Code.
11733114 Capacity: 2.5L; 220L x 160W x 105mmH
1 options
Product Code. 11733114
Brand: Gosselin™ JCP202
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For Use With: For environmental, industrial uses; Closure Material: LDPE
  • 2.5L size has molded graduations, a bowed neck and a red leakproof cap with an impervious seal
  • 5L size has a red security cap without a seal
  • 10L size has a leakproof cap with an impervious seal
TRUSTED_SUSTAINABILITY
Specifications
Capacity (English) 0.7 gal.
Capacity (Metric) 2.5 L
Certifications/Compliance EC N°1935/2004, ISO 11137
Closure Color Red
Closure Material LDPE
Closure Type Screw Cap with Wad
Description Sterile by irradiation
Diameter (Metric) Inner Neck 26 mm
Dimensions (L x W x H) 220 x 160 x 105mm
For Use With (Application) For environmental, industrial uses
Graduations Yes
Height (Metric) With Closure 105 mm
Includes Leakproof cap with an impervious seal
Length (Metric) 220 mm
Material HDPE
Product Type Jerry Can
Width (Metric) 160 mm