Learn More
Karl Hecht™ HDPE Funnels
Product Code. p-7147882
Click to view available options
Diameter (Metric)Top:
12 cm
20 cm
Unit Size:
Each
Product Code. 10688871
Brand: Karl Hecht™ 49050012
Only null left
Add to basket
Add to basket
Manufactured from HDPE for optimum strength and chemical resistance.
  • White in color
  • For general laboratory use
  • Durable polyethylene
  • Available in different diameters and stem lengths
Specifications
Diameter (Metric)Top 12 cm
Material HDPE
Color White
Length (Metric) Stem 60 mm
Product Type Funnel