Learn More
HDDC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16632-1-AP-20UL
This item is not returnable.
View return policy
Description
The function of this protein remains unknown.Specifications
| HDDC2 | |
| Polyclonal | |
| Unconjugated | |
| HDDC2 | |
| C6orf74, CGI 130, HD domain containing 2, HD domain containing protein 2, HDDC2, NS5ATP2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 361462, 51020, 69692 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3SXD3, Q7Z4H3 | |
| HDDC2 | |
| HDDC2 Fusion Protein Ag9971 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |