Description
The HDAC6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HDAC6. This antibody recognizes human antigen. The HDAC6 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
| Antigen | HDAC6 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 1C7C3 |
| Concentration | 1000 μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Quantity | 150 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 10013 |
| Show More |