Learn More
HDAC5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16166-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16885694
  • 3468.15 NOK / 150µL
Estimated Shipment: 20-12-2022
Description
This antibody only binds HDAC5. It does not cross-react with other HDACs.
HDAC5 is a member of the class II mammalian histone deacetylase family, which is structurally related to yeast HDA1. Human HDAC5 is composed of 1122 amino acid residues. The deacetylase domain of HDAC5 is located at the C-terminal half of the molecule. The N-terminal non-deacetylase domain does not show any significant homology with any published sequence.Both domains are required for HDAC5-mediated repression of gene transcription. HDAC5 interacts with a growing number of transcriptional factors including MEF2A as well as other HDAC proteins. The interacting complexes bind to specific regions of chromatin and regulate gene transcription in these regions.
Specifications
Specifications
HDAC5
Polyclonal
Unconjugated
HDAC5
Antigen NY CO 9, HD5, HDAC5, histone deacetylase 5, KIAA0600, NY CO 9
Rabbit
Antigen Affinity Chromatography
RUO
10014, 15184, 84580
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UQL6, Q9Z2V6
Hdac5
Peptide
150 μL
Primary
Rat, Mouse, Human