Learn More
HDAC4 Mouse anti-Human, Unconjugated, Clone: 1D2G12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66838-1-IG-150UL
Description
The HDAC4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HDAC4. This antibody recognizes human, rat, mouse antigen. The HDAC4 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| HDAC4 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 1D2G12 | |
| Unconjugated | |
| 150 μL | |
| 9759 | |
| IgG2a |