Learn More
HDAC4 Mouse anti-Human, Unconjugated, Clone: 1D2G12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66838-1-IG-20UL
 View more versions of this product
Product Code. 16849085
  • 1710.33 NOK / 20µL
Description
The HDAC4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HDAC4. This antibody recognizes human, rat, mouse antigen. The HDAC4 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
HDAC4
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
1D2G12
Unconjugated
20 μL
9759
IgG2a