Learn More
HDAC10 Mouse anti-Human, Unconjugated, Clone: 1A1B11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67646-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16890386
  • 4593.76 NOK / 150µL
Estimated Shipment: 16-05-2024
Description
The HDAC10 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HDAC10. This antibody recognizes human, mouse, rat antigen. The HDAC10 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
HDAC10
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, ELISA
1A1B11
Unconjugated
150 μL
83933
IgG1