Learn More
HDAC10 Mouse anti-Human, Unconjugated, Clone: 1A1B11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67646-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The HDAC10 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HDAC10. This antibody recognizes human, mouse, rat antigen. The HDAC10 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
HDAC10 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, ELISA | |
1A1B11 | |
Unconjugated | |
150 μL | |
83933 | |
IgG1 |