Learn More
HDAC1 Mouse anti-Human, Mouse, Rat, CoraLite 555, Clone: 4C4G7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL555-66085-100UL
This item is not returnable.
View return policy
Description
HDAC1 Monoclonal antibody apecifically detetects HDAC1 antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence.Specifications
| HDAC1 | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| HDAC1 | |
| HDAC1 fusion protein Ag19128 | |
| 100 μL | |
| 3065 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG1 |
| Immunofluorescence | |
| 4C4G7 | |
| CoraLite 555 | |
| BC000301 | |
| Mouse | |
| Protein G purification | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |