Learn More
HDAC1 Mouse anti-Human, Mouse, Rat, CoraLite 555, Clone: 4C4G7, Proteintech
Product Code. 17731359 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
Product Code. Quantity unitSize
17731359 20 μL 20µL
17629906 100 μL 100µL
2 options
Product Code. 17731359
Brand: Proteintech CL5556608520UL
Mouse Monoclonal Antibody
HDAC1 Monoclonal antibody apecifically detetects HDAC1 antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence.
Specifications
Antigen HDAC1
Applications Immunofluorescence
Classification Monoclonal
Clone 4C4G7
Concentration 1000 ug/mL
Conjugate CoraLite 555
Formulation PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
Gene Accession No. BC000301
Gene Symbols HDAC1
Host Species Mouse
Immunogen HDAC1 fusion protein Ag19128
Purification Method Protein G purification
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 3065
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid exposure to light.
Form Liquid
Isotype IgG1