missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hD53/TPD52L1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14732-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Four variants resulted from alternative splicing have been predicated and this antibody is expected to recognize all the variants.
This gene encodes a member of the tumor protein D52 family. The encoded protein contains a coiled-coil domain and may form homo- or hetero-dimer with TPD52 family members. The protein is reported to be involved in cell proliferation and calcium signaling. It also interacts with the mitogen-activated protein kinase kinase kinase 5 and positively regulates MAP3K5-induced apoptosis. Multiple alternatively spliced transcript variants have been observed, but the full-length nature of some variants has not yet been determined.Specifications
hD53/TPD52L1 | |
Polyclonal | |
Unconjugated | |
TPD52L1 | |
D53, hD53, TDP53, TPD52L1, TPD52L2, TPD53, tumor protein D52 like 1, Tumor protein D53 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7164 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q16890 | |
TPD52L1 | |
hD53; TPD52L1 Fusion Protein Ag6493 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |