missing translation for 'onlineSavingsMsg'
Learn More

HCK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11600-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16848783

  • 3828.53 NOK / 150µL
Estimated Shipment: 16-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

HCK is a member of the Src family of tyrosine kinases. This protein is primarily hemopoietic, particularly in cells of the myeloid and B-lymphoid lineages. It may help couple the Fc receptor to the activation of the respiratory burst. In addition, it may play a role in neutrophil migration and in the degranulation of neutrophils. Multiple isoforms with different subcellular distributions are produced dueto both alternative splicing and the use of alternative translation initiation codons, including a non-AUG (CUG) codon.
Specifications

Specifications

HCK
Polyclonal
Unconjugated
HCK
HCK, hemopoietic cell kinase, JTK9, p59 HCK/p60 HCK, Tyrosine protein kinase HCK
Rabbit
Antigen Affinity Chromatography
RUO
15162, 25734, 3055
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P08103, P08631, P50545
Hck
HCK Fusion Protein Ag2132
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers