missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ HCAP-G (Human) Recombinant Protein
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Description
- Sequence: TTFQNEDEKNKEVYMTPLRGVKATQASKSTQLKTNRGQRKVTVSARTNRRCQTAEADSESDHEVPEPESEMKMRLPRRAKTAALEKSKLNLAQFLNEDLS
Specifications
Specifications
| Accession Number | AAH00827 |
| Gene ID (Entrez) | 64151 |
| Name | non-SMC condensin I complex, subunit G |
| Preparation Method | Wheat germ expression system |
| Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
| Quantity | 10 μg |
| Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Gene Alias | CAPG, CHCG, FLJ12450, HCAP-G, MGC126525, NY-MEL-3 |
| Gene Symbol | NCAPG |
| Species | Wheat Germ (in vitro) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction